Z505846
Nalgene® quick filling/venting closure
fits tubing, 3/8 in., number of ports, 2
Synonym(e):
transfer cap, vented cap
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.35
Empfohlene Produkte
Leistungsmerkmale
fits tubing 3/8 in.
number of ports 2
Verpackung
pack of 1 ea
Hersteller/Markenname
Nalgene 2158-0022
Verwandte Kategorien
Allgemeine Beschreibung
Available in two and three port configurations, and is ideal for filling/dispensing into and out of vacuum trap systems with carboys and bottles. Quick disconnect ports fit either 1/4 in. or 3/8 in. I.D. tubing, and have a built-in shut off valve if tubing inserts are taken out. Fits onto all carboys and bottles with 83 mm neck. Leakproof.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Zubehör
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z505846-1EA | 4061832618197 |