Merck
Z285072
Sigma-Aldrich
Glas-Col® Series O extension support
for flask size, 250 ‑ 300 mL
About This Item
UNSPSC-Code:
41102405
NACRES:
NB.76
Materialien
steel
Leistungsmerkmale
for flask size 250 ‑ 300 mL
Hersteller/Markenname
Glas-Col 100E0525
Allgemeine Beschreibung
The steel support rings with fabric strap basket hold spherical and hemispherical mantles. Attach to lattice systems or ring stands.
Rechtliche Hinweise
Glas-Col is a registered trademark of Glas-Col LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)