Merck
Z280380
Sigma-Aldrich
PVC-Laborschlauch
I.D. × O.D. 1/4 in. × 1/2 in.
Synonym(e):
Saint Gobain Versilon ABH02019
About This Item
UNSPSC-Code:
41105108
NACRES:
NB.51
Verpackung
pkg of 100 feet
ID × AD
1/4 in. × 1/2 in.
Wandabmessungen
1/8 in.
Allgemeine Beschreibung
General-purpose polyvinyl chloride laboratory tubing. Clear, flexible tubing ideal for drains and rinse tubes. Max. temp.: 165°F (73.4°C).
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)