Merck
WHAWB120028
QIAcard FTA CloneSaver Cards
96 well format, pkg of 5 cards
Synonym(e):
QIAcard FTA CloneSaver (5), Whatman FTA, Z747440
About This Item
UNSPSC-Code:
41105500
NACRES:
NB.22
Beschreibung
CloneSaver Card, 96 well format, 5 μL maximum volume/sample area, 480 μL maximum total volume/card
Hersteller/Markenname
Qiagen WB120028
Verpackung
pkg of 5 cards
Allgemeine Beschreibung
CloneSaver Card is designed for the collection, long-term storage, and purification of plasmid and BAC DNA from bacterial clones in a 96-well format
Leistungsmerkmale und Vorteile
  • Captures and stabilizes bacterial artificial chromosome (BAC) and plasmid DNA in one easy step: without the requirement of reagents, centrifuges or other materials
  • Serves as a secure, reliable alternative to glycerol stocks
  • Simplifies storage, transport and access to clone libraries: conveniently, easily and inexpensively
  • Ensures faster clone identification and recovery through sample ID chart, instruction for use and area for bar code labeling
  • Produces ready-to-use vector DNA in less than 30 minutes: polymerase chain reaction (PCR) ready bacterial clones
  • Protects clone stocks from phage infections by inactivating bacteria and phage
Sonstige Hinweise
Field of Use : For internal research use only
Rechtliche Hinweise
FTA is a trademark of Qiagen Group
QIAcard is a trademark of Qiagen Group
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Artikel
This article describes the evaluation of Whatman FTA cards from GE Healthcare for their ability to collect, store, and isolate high-quality RNA from a variety of crude biological samples.
Polymerase chain reaction (PCR) is a technique that results in exponential amplification of a target DNA sequence.
Protokolle
This study examined the yield and quality of DNA from samples applied to Whatman FTA cards, using five common methods of DNA extraction.
This page shows options for sample collection, transport, archiving, and DNA purification.
Nucleic Acid Sample Preparation Methodology