Merck
WHA10451710
Whatman® pressure filtration devices
MD 142/7/3 PTFE filter holder, 1500 mL infusion vessel, capacity 142 mm
Synonym(e):
Z751294
About This Item
UNSPSC-Code:
41104900
NACRES:
NB.24
Materialien
PTFE housing
Leistungsmerkmale
1500 mL infusion vessel
Verpackung
pk of 1
Hersteller/Markenname
Whatman 10451710
Whatman Article No. 28415695 (US reference)
Kapazität
142 mm
Allgemeine Beschreibung
Pressure filtration devices with a sample loading cylinder are suitable for batch filtration of samples from 20 ml. Devices without infusion cylinders are connected inline and are suitable for larger volumes of several liters.
Filtration of liquids and gases is possible, including sterile filtration* of serums or clear filtration of media that are difficult to filter, especially those that are highly viscous.
Leistungsmerkmale und Vorteile
  • Membranes, paper or glass fiber filter discs can be used.
  • Cleaning and changing of filters is completed in a few steps.
Sonstige Hinweise
Field of Use: For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)