WHA10370010
Whatman® glass microfiber filters with inorganic binder, Grade GF 6
diam. 185 mm
Synonym(e):
Z749893
Alle Fotos(1)
About This Item
UNSPSC-Code:
41104900
NACRES:
NB.24
Leistungsmerkmale
hydrophilic
Verpackung
pk of 100
Hersteller/Markenname
Whatman 10370010
Whatman Article No. 28414750 (US reference)
Durchmesser
185 mm
Allgemeine Beschreibung
Good retention for very fine particles. This filter is used in water pollution applications, for removing protein from difficult-to-filter beers, for determination of chlorophyll and phytoplankton residues, for the determination of filterable substances and the residue on ignition (dry weight), for the analysis of aggressive media (e.g., acidic gases), for scintillation measurements and for determination of the elemental iron content in the presence of iron oxides.
Sonstige Hinweise
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)