TEM-FCF100NI50
Formvar/Carbon Supported Nickel Grids
size 100 mesh, box of 50 ×
Synonym(e):
Formvar/Carbon 100 Mesh, Ni, 50pk
Alle Fotos(1)
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Empfohlene Produkte
Verpackung
box of 50 ×
Größe
100 mesh
Verwandte Kategorien
Allgemeine Beschreibung
A formvar coated TEM grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections.
Thickness: 10nm Formvar and 1nm Carbon
Thickness: 10nm Formvar and 1nm Carbon
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)