Merck
TEM-CFT300CU
Sigma-Aldrich
Carbon Film Supported Copper Thin Bar Square Mesh Grid
size 300 mesh, box of 25
Synonym(e):
CARBON FILM ONLY ON THIN BAR 300 MESH COPPER GRIDS 25/BX
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Verpackung
box of 25
Größe
300 mesh
Allgemeine Beschreibung
A 5-6 nm thin film of pure carbon supported thin bar square mesh grid. Copper grid, 300 mesh. Carbon film show better electrical and heat conduction in the electron beam.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)