Merck
TEM-CF400CU
Sigma-Aldrich
Carbon Film Supported Copper Square Mesh
size 400 mesh, box of 25
Synonym(e):
CARBON FILM 400# COPPER 25/PK
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Verpackung
box of 25
Größe
400 mesh
Allgemeine Beschreibung
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Copper grid, 400 mesh.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)