SYNS391656
Synthware™ high vacuum double oblique glass stopcock
bore 4 mm, stem O.D. 10 mm, Plug Size: 16.2/56
Synonym(e):
Synthware S391656
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
S391656
Bohrung
4 mm
Schaft AD
10 mm
Verwandte Kategorien
Allgemeine Beschreibung
Synthware™ glass stopcock features solid glass plug. Barrel and plug are precision ground with corresponding numbers on the barrel and plug. With constant pressure retention spring and clip.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)