SYNP150050M
Synthware™ synthesis vessel with lower joint and PTFE T-bore stopcock
50 mL, joint: ST/NS 24/40, porosity: medium, with drip tip
Synonym(e):
Synthware P150050M, Solid phase peptide synthesis vessel
Alle Fotos(1)
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.51
Empfohlene Produkte
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware P150050M
Kapazität
50 mL
50 mL
Verwandte Kategorien
Allgemeine Beschreibung
Solid phase peptide synthesis vessels feature 2 mm PTFE stopcocks and fritted glass supports for resin beads during operation. The 24/40 inner drip joint can be assembled with other standard glassware. The vessel has a 8 mm hose connection for vacuum and the stopcock has an 8 mm O.D. sidearm for inert gas introduction. Vessels are supplied with PTFE-faced silicone lined Nylon cap.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)