SYNP130100M
Synthware™ peptide synthesis vessel with fritted disc and T-bore PTFE stopcock
100 mL, porosity: medium, GL 25
Synonym(e):
Synthware P130100M, Solid phase peptide synthesis vessel
Alle Fotos(1)
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.51
Empfohlene Produkte
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware P130100M
Kapazität
100 mL
100 mL
Fitting
GL 25
Verwandte Kategorien
Allgemeine Beschreibung
With a fritted disc of coarse or medium porosity to support resin. Top of the vessel has GL thread and a PTFE lined screw cap. PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)