Merck
SYNF984500
Synthware Kuderna-Danish flask
capacity 500 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 19/22
Synonym(e):
Synthware F984500
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
borosilicate glass flask
pear-shaped
single-neck
Verbindung
bottom joint: ST/NS19/22
joint: ST/NS top24/40
Agentur
suitable for EPA 1613
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F984500
Kapazität
500 mL
Allgemeine Beschreibung
Top joint is a 24/40 outer while the lower joint is a 19/22 inner.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)