Merck
SYNF902450H
Synthware pear-shaped Schlenk flask with stopcock
capacity 50 mL, joint: ST/NS 24/40
Synonym(e):
Synthware F902450H, modified Schlenk reaction flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
borosilicate glass flask
pear-shaped
single-neck
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F902450H
Kapazität
50 mL
Allgemeine Beschreibung
A suffix of G indicates a glass stopcock, H indicates a high-vacuum valve, no letter suffix indicates a PTFE Stopcock.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)