SYNF791100
Synthware™ modified evaporation flask
capacity 100 mL, neck joint: ST/NS 14/20
Synonym(e):
Synthware F791100, rotary evaporator flask
Empfohlene Produkte
Materialien
glass flask
round bottom flask
Verbindung
joint: ST/NS neck14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F791100
Kapazität
100 mL
Verwandte Kategorien
Allgemeine Beschreibung
Low profile design sits upright on bench and also provides for easy product access with a spatula. The unique flask shape is excellent for the controlled evaporation of foaming solvents such as toluene or xylene on a rotary evaporator
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)