Merck
SYNF764108
Synthware storage tube with standard taper side arm
capacity 50 mL, joint: ST/NS 24/40, valve size 0-8 mm
Synonym(e):
Synthware F764108, storage tube with outer joint
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Materialien
black cap
cap (plug seal)
colorless tube
glass tube
round bottom
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F764108
Kapazität
50 mL
Ventilgröße
0-8 mm
Allgemeine Beschreibung
Supplied with high vacuum valve with PTFE O-ring at tip.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)