Merck
SYNF341200
Synthware two neck pear shaped flask
200 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Synonym(e):
Synthware F341200, 2-neck pear shape flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
borosilicate glass flask
pear-shaped
two-neck
Verbindung
center joint: ST/NS14/20
side joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F341200
Kapazität
200 mL
200 mL
Allgemeine Beschreibung
Pear shape with joints. Heavy wall design.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)