Merck
SYNC564175
Synthware coil-style reflux condenser with removable hose connections
joint: ST/NS 24/40, hose size 10 mm
Synonym(e):
Synthware C564175, coil-type condenser
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware C564175
Schlauchgröße
10 mm
Allgemeine Beschreibung
Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with volatile solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)