SYNA841420
Synthware™ vacuum filtration adapter
bottom joint: ST/NS 14/20, Stopper Size: #1, stopper size #1
Synonym(e):
Synthware A841420, Glass adapter vacuum filtration
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Empfohlene Produkte
Verbindung
bottom joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
A841420
Synthware A841420
Allgemeine Beschreibung
Used with plain stem Buchner funnels and pluro-stopper. Hose connection has an O.D of 10 mm at the largest serration. Order pluro-stoppers below separately.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)