SYNA692440
Synthware™ 90 degree vacuum adapter with glass stopcock
female joint: ST/NS 24/40
Synonym(e):
Synthware A692440, Flow control adapter
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Empfohlene Produkte
Verbindung
female joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A692440
Verwandte Kategorien
Allgemeine Beschreibung
For regulating the input of gases or liquids into reaction systems. With 8 mm or 10 mm hose connection. With 8 mm or 10 mm hose connection.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
SYNA692440-1EA | 4061832189659 |