SYNA291922
Synthware™ vacuum or argon adapter with hose connection on side
joint: ST/NS 19/22
Synonym(e):
Synthware A291922, inert gas adapter, vacuum adapter
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Empfohlene Produkte
Verbindung
joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A291922
Allgemeine Beschreibung
With hose connection on the side and male joint.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)