SLW1519/06
Pyrex® Media-Lab Bottles, amber glass, with-out cap and pouring ring, with printed trace code
capacity 250 mL
Synonym(e):
Pyrex, SciLabware 1519/06
About This Item
Empfohlene Produkte
Materialien
round amber glass bottle
Verbindung
Joints threaded neck (GL45)
Sterilität
non-sterile
Leistungsmerkmale
graduations
without cap
with printed trace code
without pouring ring
Verpackung
pack of 10 ea
Hersteller/Markenname
SciLabware 1519/06
Höhe
138 mm
Grundkörperdurchm.
70 mm
Kapazität
250 mL
Allgemeine Beschreibung
- Bottles only. Supplied without cap or pouring ring
- Complies with ISO 4796
- Manufactured from amber-coated borosilicate glass
- UV absorbent up to 500 nm
- Ideal for storage of light sensitive materials
- Printed with Trace Code for downloadable batch certificate
- Batch certificate available to download from www.scilabware.com
Leistungsmerkmale und Vorteile
- Bottles only. Supplied without cap or pouring ring
- Complies with ISO 4796
- Manufactured from amber-coated borosilicate glass
- UV absorbent up to 500 nm
- Ideal for storage of light sensitive materials
- Printed with Trace Code for downloadable batch certificate
- The 25 ml size bottle features a glass lip and does not require a pouring ring.
- Never autoclave bottles with a fully tightened screw cap. Always slacken off. Failure to follow this procedure can lead to breakage of the bottle.
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
SLW1519/06-10EA | 4061832437880 |