SLW1060/10D
Pyrex® Watch glasses
nominal diameter 80 mm
Synonym(e):
Pyrex, SciLabware 1060/10D
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121801
NACRES:
NB.43
Empfohlene Produkte
Verpackung
pkg of 10
Hersteller/Markenname
1060/10D
SciLabware
Nenndurchmesser
80 mm
Verwandte Kategorien
Allgemeine Beschreibung
Watch glasses
- For evaporation work and covering heated beakers
- Manufactured from heat resisting Pyrex borosilicate glass
- Fire-polished edges to reduce chipping and breakage
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)