Merck
SLW1000/18D
Pyrex® Griffin beakers, low form, with printed trace code
600 mL
Synonym(e):
Pyrex, SciLabware 1000/18D
About This Item
UNSPSC-Code:
41121803
NACRES:
NB.43
Materialien
borosilicate glass
white graduation
Leistungsmerkmale
low form
with printed trace code
Hersteller/Markenname
1000/18D
SciLabware
H × Durchm.
125 mm × 90 mm
Kapazität
600 mL
Allgemeine Beschreibung
  • Complies with ISO 3819 and DIN 12331
  • Manufactured from Pyrex borosilicate glass
  • Excellent thermal performance
  • High resistance to chemical attack
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • White graduations and marking spot
Sonstige Hinweise
Browse all glass beakers
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)