Merck
S977551
9-{[3-(9H-CARBAZOL-9-YLMETHYLENE)-2,4-DIPHENYLCYCLOBUTYLIDENE]METHYL}-9H-CARBAZOLE
AldrichCPR
About This Item
Lineare Formel:
C42H30N2
CAS-Nummer:
Molekulargewicht:
562.721
MDL-Nummer:
Sonstige Hinweise
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
S977551-1EA