MT51107687
Mettler Toledo DX series ISE half-cell membrane kit
For Lithium ISE
Alle Fotos(1)
About This Item
UNSPSC-Code:
39121436
NACRES:
NB.79
Hersteller/Markenname
Mettler-Toledo 51107687
Allgemeine Beschreibung
DX series ion specific membrane kit. Mount the new membrane onto the shaft of your ISE half-cell, and you have a new ISE!
Kit contains one membrane plus electrolyte
Kit contains one membrane plus electrolyte
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)