Merck
GF41816662
Stainless steel - 17-4ph
rod, Fe/Cr17%/Ni 4%, 8.0 mm diameter, length 200 mm
Synonym(e):
FL017910
About This Item
UNSPSC-Code:
12352200
NACRES:
NA.23
Form
rod
Hersteller/Markenname
Goodfellow 418-166-62
Länge
200 mm
Durchmesser
8.0 mm