Wichtige Dokumente
DWK73500-650
KIMBLE® disposable rimless culture tube
round bottom borosilicate glass, tube diam. (6 mm) (50 mm), non-sterile
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121706
NACRES:
NB.11
Empfohlene Produkte
Materialien
round bottom borosilicate glass
Sterilität
non-sterile
Verpackung
case of 1000 ea shrink-wrapped trays
Hersteller/Markenname
Kimble® 73500-650
Röhrchendurchm.
(6 mm) (50 mm)
Allgemeine Beschreibung
Plain Disposable Borosilicate Glass Tubes
- These premium quality tubes feature sturdy, uniform bottoms and consistent lengths.
- 51 expansion borosilicate glass offers excellent chemical resistance against sodium leaching, a factor common in soda lime glass products
- Tubes are packed in convenient trays for ease of use on bench tops and in drawers
- Trays are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-transit breakage and contamination
- No marking spot
- Designed from ASTM Specification E890
- Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Leistungsmerkmale und Vorteile
- Complies with ASTM Specification E890
- Manufactured from borosilicate glass 5.1
- 51 expansion borosilicate glass offers excellent chemical resistance against sodium leaching, a factor common in soda lime glass products
- Tubes are packed in convenient trays for ease of use on bench tops and in drawers
- The 6 x 50 mm size is often referred to as a "Durham tube"
- Trays are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-transit breakage and contamination
- No marking spot
Rechtliche Hinweise
KIMBLE is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)