Merck
CLS4320C5L
Pyrex® round-bottom heavy wall boiling flask, short neck with joint
capacity 5,000 mL, joint: ST/NS 45/50
Synonym(e):
bottom flask, flask round, round flask, Corning® 4320C
About This Item
UNSPSC-Code:
41121804
Materialien
borosilicate glass
round bottom flask
short
Verbindung
joint: ST/NS45/50
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard taper joint
Verpackung
case of 1
Hersteller/Markenname
Corning 4320C-5L
Kapazität
5,000 mL
Halsumfang
(1)
Allgemeine Beschreibung
Pyrex heavy duty boiling flask provides increased mechanical strength. The flask features walls that are 25-30% thicker than the standard version.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)