Merck
CLS4100125
Pyrex® flat-bottom boiling flask, short neck with 24/40 joint
capacity 125 mL
Synonym(e):
boiling flask, flask glass, flat bottom flasks, Corning® 4100
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass
flat bottom flask
short
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper Joint
Verpackung
case of 12
Hersteller/Markenname
Corning 4100-125
Kapazität
125 mL
Halsumfang
(1)
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)