Merck
CLS2962300
Pyrex® hydrometer jar
capacity 300 mL
Synonym(e):
Corning® 2962
About This Item
UNSPSC-Code:
41113024
NACRES:
NB.43
Hersteller/Markenname
Corning 2962-300
AD × H
39 mm × 381 mm
Kapazität
300 mL
Allgemeine Beschreibung
Breakage is minimized by the heavy wall construction. The large hexagonal base, sealed to the cylinder body, increases stability. Meets ASTM specifications D-287 and E-100.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)