CAD6512
1-way tubing connector
MLL to hose end for 1/4 in. to 5/16 in. I.D. tubing (316SS)
Synonym(e):
Tubing Connector
Alle Fotos(1)
About This Item
UNSPSC-Code:
40183110
NACRES:
NB.03
Empfohlene Produkte
Verpackung
pack of 5 ea
Hersteller/Markenname
Cadence Science 6512
Verwandte Kategorien
Allgemeine Beschreibung
- Plated brass or 316 stainless steel
- Meets ISO 594 specifications
- Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
Micro-Mate® Female Luer Lock (MMFLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.
Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.
Male Luer (ML) - male Luer slip that fits with a female Luer.
Rechtliche Hinweise
Micro-Mate is a registered trademark of Cadence Science
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
CAD6512-5EA | 4061832517810 |