Merck
CAD4002
Standard point hypodermic needles, 304 stainless steel
gauge 27, L 1/2 in., nickel plated hub
About This Item
UNSPSC-Code:
42142523
NACRES:
NB.03
Materialien
304 stainless steel needle
nickel plated hub
Verpackung
pack of 12 ea
Hersteller/Markenname
Cadence Science 4002
Länge
1/2 in.
Manometer
27
Allgemeine Beschreibung
Hypodermic needles are precision made and designed to ensure strict adherence with U.S. Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannula are fabricated of type 304 stainless steel tubing which must pass rigorous quality control prior to acceptance and use.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)