BR90629
BRAND® glass beaker with spout, tall form
volume 150 mL, with graduation
Synonym(e):
Beakers, Glass
Alle Fotos(2)
About This Item
UNSPSC-Code:
41121803
NACRES:
NB.43
Empfohlene Produkte
Materialien
borosilicate glass 3.3
Leistungsmerkmale
with graduation
Hersteller/Markenname
BRAND 90629
Volumen
150 mL
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® Glass Beaker with Spout, tall form, is constructed from high-quality Boro 3.3 glass, ensuring durability and reliability in laboratory environments. Compliant with DIN EN ISO 3819 standards, this beaker features clear graduation markings for accurate liquid measurement. The tall form design provides increased capacity, while the integrated spout facilitates easy pouring, minimizing spills and enhancing precision during liquid transfer.
Sonstige Hinweise
Browse all glass beakers
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR90629-10EA | 4061831861563 |