Merck
BR90154
BRAND® Griffin beaker with spout
low form, PFA, volume 500 mL
Synonym(e):
Beakers, PFA, Plastics
About This Item
UNSPSC-Code:
41121803
NACRES:
NB.43
Materialien
PFA
Leistungsmerkmale
graduations
low form
Hersteller/Markenname
BRAND 90154
Volumen
500 mL
Allgemeine Beschreibung
The BRAND® Griffin Beaker with Spout is crafted from high-quality ETFE, offering excellent chemical resistance and durability for laboratory applications. This translucent beaker features clear graduation markings for precise measurement, allowing for accurate liquid handling. The beaded rim enhances strength and provides a comfortable grip, while the integrated spout facilitates easy pouring, minimizing spills and improving accuracy during liquid transfer. Ideal for a variety of experiments, this beaker combines functionality with reliability, making it an essential tool for any laboratory setting.
Sonstige Hinweise
Browse all plastic beakers
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)