Merck
BR781334
BRAND® strips of 8 caps
polypropylene, clear, flat
Synonym(e):
PCR, PCR Tubes, Tube Strips, Tubes
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
Materialien
clear
flat
polypropylene
Verpackung
pack of 125 ea
Hersteller/Markenname
BRAND 781334
Allgemeine Beschreibung
With PCR cap strips you can seal PCR strips and plates with 24, 48, and 96 wells reliably and protect your samples effectively against evaporation and contamination.The shape of the cap strips is optimally matched to BRAND PCR tube strips and plates so they are easy to open and close. To prevent contamination, the cap strips are equipped with two side tabs for exact positioning before closing and a small opening attachment on each cap. This prevents contact with the inside of the cap.The highly transparent flat cap strips are ideal for qPCR because the cap shape allows accurate fluorescence measurement. With domed cap strips, thermal cyclers without pressure sensors exert a stronger force, thus further increasing the sealing effect.Manufactured under cleanroom conditions. BIO-CERT® PCR QUALITY: Free of human DNA, RNase, DNase, Pyrogens and PCR-Inhibitors
Leistungsmerkmale und Vorteile
  • Effective protection against evaporation and contamination
  • The right cap shape for your application
  • Easy to open and close without risk of contamination
Rechtliche Hinweise
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)