Merck
BR708728
BRAND® disposable BLAUBRAND® micropipettes, intraMark
tube volume 20-40 μL , Dual marks, red
Synonym(e):
Micropipettes, Pipettes, intraMARK
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Materialien
red
Leistungsmerkmale
DE-M marking, To Contain
Verpackung
pack of 1000 ea (4 boxes of 250)
Hersteller/Markenname
BRAND 708728
Länge
125 mm
Röhrchenvolumen
20-40 μL , Dual marks
Allgemeine Beschreibung
DIN ISO 7550. DE-M marking for volumes greater than 5 µl. Calibrated to contain (TC, In). Color code to ISO for clear identification.
Rechtliche Hinweise
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)