BR707535
BRAND® dispenser cylinder with valve block for Titrette®
for 25 mL from serial-number 01K
Synonym(e):
Accessories, Bottle-top Burette, Burettes, Titrette®
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 707535
Verwandte Kategorien
Allgemeine Beschreibung
Structural change from serial-number 01K (January 2012).The assembly of the titrating tube to the valve block has been changed. When ordering spare parts, the serial number must be accounted for.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Titrette is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR707535-1EA | 4061831866773 |