BR462026
BRAND® rubber sleeve for filter crucibles
O.D. 41 mm
Synonym(e):
Filter, Rubber sleeve
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.13
Empfohlene Produkte
Materialien
EPDM rubber
Verpackung
pack of 10 ea
Hersteller/Markenname
BRAND 462026
AD
41 mm
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® Rubber Sleeve for Filter Crucibles is made from high-quality EPDM rubber, designed to provide a secure and reliable fit for various filter crucibles according to the identification code. This durable sleeve enhances the functionality of filter crucibles by ensuring a tight seal, preventing leaks and contamination during filtration processes.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR462026-10EA | 4061831856903 |