Merck
BR4600170
BRAND® Dispensette® S Analog-adjustable bottle-top dispenser
volume range 10-100 mL, without recirculation valve
Synonym(e):
bottle top dispensers
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Leistungsmerkmale
incremental 1 mL
DE-M marking
without recirculation valve
Hersteller/Markenname
BRAND 4600170
Volumenbereich
10-100 mL
Fitting
(GL 32-33)
GL 38
S 40
Allgemeine Beschreibung
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Leistungsmerkmale und Vorteile
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Sonstige Hinweise
Analog-adjustable - Rapid and safe volume setting with new internal scalloped track. Simple calibration adjustment with a supplied tool.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Protokolle
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.