Merck
BR43208
BRAND® density bottle, Gay-Lussac pattern, uncalibrated
capacity 10 mL, borosilicate glass
Synonym(e):
Gay-Lussac
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.71
Materialien
borosilicate glass
glass stopper
narrow-mouth bottle
round bottle (Gay-Lussac)
white bottle
Verbindung
joint ST/NS neck10/19
Leistungsmerkmale
with cap
Verpackung
pack of 2 ea
Hersteller/Markenname
BRAND 43208
Kapazität
10 mL
Allgemeine Beschreibung
DIN ISO 3507, type Gay-Lussac. Stopper NS 10/19 with capillary. Top of stopper ground and polished. Nominal capacity printed on the bottom.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Protokolle
How to Use a Density Bottle
Global Trade Item Number
SKUGTIN
BR43208-2EA4061831856491