BR29125
BRAND® pipette rinsing system, rinser for pipette
high-density polyethylene, capacity 17 L, L × I.D. 1000 mm × 150 mm
Synonym(e):
Pipette Rinsing Systems, Pipette rinser, Pipettes
Alle Fotos(1)
About This Item
UNSPSC-Code:
41103200
NACRES:
NB.35
Empfohlene Produkte
Materialien
high-density polyethylene
Hersteller/Markenname
BRAND 29125
L × ID
1000 mm × 150 mm
Kapazität
17 L
Verwandte Kategorien
Allgemeine Beschreibung
PE-HD. Trouble-free and gentle cleaning of pipettes with repetitive siphon rinsing. Water supply connection with hose nozzle outer Ø 12 mm. Soaking jar and pipette basket sold separately. Adjust the water flow to approx. 2 L/min.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR29125-1EA | 4061831865110 |