Merck
BR152810
BRAND® tubing connector, T-shape
polypropylene, for tubing i.d., 8 ‑ 9 mm
Synonym(e):
Tubing, Tubing Connectors
About This Item
UNSPSC-Code:
40183110
NACRES:
NB.43
Materialien
polypropylene
Leistungsmerkmale
for tubing i.d. 8 ‑ 9 mm
Verpackung
pack of 20 ea
Hersteller/Markenname
BRAND 152810
Allgemeine Beschreibung
The BRAND® T-shaped tubing connector is constructed from translucent polypropylene (PP), allowing for easy visibility of fluid flow. This connector is designed to create a branching point in tubing systems, enabling the connection of multiple tubes at different angles. Its durable construction ensures a secure and reliable fit, minimizing the risk of leaks or disconnections during operation.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)