Merck
BR144515
BRAND® conical joint stopper, borosilicate glass
joint: ST/NS 12/21, solid
Synonym(e):
Conical joint stoppers, Stoppers
About This Item
UNSPSC-Code:
41122500
NACRES:
NB.51
Materialien
glass
octagonal borosilicate glass (grip)
Verbindung
joint: ST/NS12/21
Beschreibung
octagonal grip
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 144515
Allgemeine Beschreibung
DIN 12252. Solid or semi-hollow, with octagonal grip.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)