BMSV0020QD
Aspire™ Laboratory Aspirator Accessories
Option quick disconnect kit, includes 2 male/female attachments & silicone tubing
Alle Fotos(1)
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.79
Empfohlene Produkte
Hersteller/Markenname
Benchmark Scientific V0020-QD
Verwandte Kategorien
Allgemeine Beschreibung
Accessories designed for use with Aspire laboratory aspirator.
Quick disconnect kit comprises of 2 male/female attachments & silicon tubing and enables swift changes between aspiration tools.
Rechtliche Hinweise
6538 is a trademark of American Type Culture Collection
Aspire is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)