Merck
BAH369460010
SP Bel-Art® Sterileware Long Handled Sampling Spoon
white polystyrene, nominal capacity 14.79 mL , 3tsp, L × W 17.8 cm (7 in.) × 1.5625 in. (40 mm), sterile, pack of 10 ea (individually wrapped)
Synonym(e):
Lab Scoop, dispensing tool, spoon
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.35
Materialien
white polystyrene
Sterilität
sterile
Verpackung
pack of 10 ea (individually wrapped)
Hersteller/Markenname
Bel-Art H36946-0010
L × B
17.8 cm (7 in.) × 1.5625 in. (40 mm)
Nennkapazität
14.79 mL , 3tsp
Allgemeine Beschreibung
Designed to gather small amounts of sample material, especially in hard to reach containers.
Leistungsmerkmale und Vorteile
  • Long handle reaches into jars, bottles and other confined areas
  • Minimizes the potential of personal contact with sampled material
  • Sterile, FDA grade polystyrene safe for food, drug and cosmetic use
  • Individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing
Note: Sterilized products are not returnable.
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)