BAH189620002
SP Bel-Art® Poxygrid® Wall mounted Microliter Pipettor Rack
to hold, Gilson Pipetman
Synonym(e):
pipette wall mounting rack
Alle Fotos(3)
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.43
Empfohlene Produkte
Leistungsmerkmale
to hold Gilson Pipetman
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art H18962-0002
L × B × H
9 in. (229 mm) × 3.875 in. (98 mm) × 8 in. (203 mm)
Verwandte Kategorien
Allgemeine Beschreibung
Choose from 3 styles of epoxy-coated aluminum rack to hold Gilson Pipetman® pipettors.
Leistungsmerkmale und Vorteile
- Provides storage and easy access for up to 10 pipettors plus up to two Pipet-Aid® pipettes
- H18962-0001 for desktop storage
- H18962-0002 for wall mounting; has two holes for screw attachment to the wall and double faced tape (included)
- H18962-0004 has extended base with raised lip to hold plastic tip boxes; suitable for wall mounting or for free standing on a bench
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Pipet-Aid is a registered trademark of Drummond Scientific Co.
Pipetman is a registered trademark of Gilson S.A.S. LLC
Poxygrid is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)