Merck
BAF378740005
SP Bel-Art® Sterileware Powder Thief
capacity × L 5 mL (0.17 oz) × 19.75 in. (50 cm), pack of 20 ea (individually wrapped)
Synonym(e):
sampler
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.35
Materialien
high-density polyethylene
Verpackung
pack of 20 ea (individually wrapped)
Hersteller/Markenname
Bel-Art F37874-0005
capacity × L
5 mL (0.17 oz) × 19.75 in. (50 cm)
Allgemeine Beschreibung
Available in two lengths each with four tip volumes, these sterile powder thieves easily sample from targeted locations within bulk powders. Used by pharmaceutical, food, cosmetic and chemical companies.
Leistungsmerkmale und Vorteile
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) thief heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10^-6)Ready for one time use and disposal
  • Lot stamped for accurate tracing
Note: Sterilized products are not returnable.
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)