BAF371250014
Spinfin® magnetic stirring bar
H × diam. 0.375 in. (9.5 mm) × 0.3125 in. (7.9 mm)
Synonym(e):
Spinfin® magnetic stirring bar
Alle Fotos(4)
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.51
Empfohlene Produkte
Materialien
PTFE-coated
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F37125-0014
H × Durchm.
0.375 in. (9.5 mm) × 0.3125 in. (7.9 mm)
Verwandte Kategorien
Allgemeine Beschreibung
Mit dünnen Flossen oben
Spinfin®magnetic stirring bars agitate liquids in round bottom vessels such as test tubes, cylinders and flasks, where other stirring bars cannot be used.
Leistungsmerkmale und Vorteile
- Maintains its position in a vessel and will not wobble
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating – Superior Chemical Compatibility
- Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
- Autoclave safe
Rechtliche Hinweise
Spinfin is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)