Merck
BAF371220045
Spinbar® Magnetrührstäbchen
polygon white PTFE-coated, L × diam. 1.75 in. (45 mm) × 0.3125 in. (8 mm)
Synonym(e):
Spinbar® magnetic stir bar
About This Item
UNSPSC-Code:
41122800
Materialien
polygon white PTFE-coated
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F37122-0045
L × Durchm.
1.75 in. (45 mm) × 0.3125 in. (8 mm)
Allgemeine Beschreibung
  • Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
  • Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
  • Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
  • Autoclave safe.
These stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
Leistungsmerkmale und Vorteile
  • Center molded pivot ring minimizes the contact area between the bar and the vessel; reduces friction and chattering
  • Marring of plastic containers is diminished
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE® brand PTFE Coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Spinbar is a registered trademark of SP Scienceware, a division of SP
Teflon is a registered trademark of E. I. du Pont de Nemours and Company